Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KSHV ORF62 Sheep anti-Virus, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody
$243.50 - $487.50
Specifications
Antigen | KSHV ORF62 |
---|---|
Dilution | Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KSHV ORF62 Polyclonal antibody specifically detects KSHV ORF62 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot BlotSpecifications
KSHV ORF62 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot | |
Unconjugated | |
Sheep | |
Virus | |
Kaposi's Sarcoma Virus, Kaposi's sarcoma-associated herpesvirus, ORF62 | |
A fusion protein that contains at its amino terminus the maltose binding protein and at its carboxy terminus the protein encoded by the open reading frame 62 (ORF62) (Uniprot: Q2HR66) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
4961461 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title