Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KSHV ORF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519852 Supplier Invitrogen™ Supplier No. PA519852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-19852 targets KSHV ORF8 in indirect ELISA, WB applications and shows reactivity with Human samples. The PA5-19852 immunogen is synthetic peptide, corresponding to amino acids sequence of KSHV ORF8. .

Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. KSHV ORF8 is a virion envelope-associated protein. Akula et al (2003) report it acts as a Glycoprotein B mediated signaling during virus entry. This Glycoprotein possesses the Arg-Gly-Asp (RGD) motif and utilizes alpha3beta1 integrin as one of the receptors for its entry into the target cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene Alias Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids sequence of KSHV ORF8.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.