Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KSR1 Antibody (SR1095), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | KSR1 |
---|---|
Clone | SR1095 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
KSR1 Monoclonal antibody specifically detects KSR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
KSR1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
kinase suppressor of ras, kinase suppressor of ras 1, KSRRSU2 | |
A synthesized peptide derived from human KSR1 (Uniprot #: Q8IVT5) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1095 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8844 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title