Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KSR1, Rabbit anti-Human, Clone: 3A4, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18922
Specifications
KSR1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide). | |
KSR/RSU2 | |
Rabbit | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
3A4 | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
kinase suppressor of ras 1 | |
KSR1 | |
A synthetic phosphopeptide corresponding to residues surrounding S392 of human KSR1. | |
RUO | |
8844 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction