Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KSV Cyclin D Monoclonal Antibody (94B)
GREENER_CHOICE

Catalog No. PIMA191431
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191431 Supplier Invitrogen™ Supplier No. MA191431
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MA1-91431 detects D type cyclin from virus samples.

This protein was identified by the interaction with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein was shown to interact with Cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSV Cyclin D
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 94B
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias B-cell CLL/lymphoma 1; B-cell lymphoma 1 protein; BCL1; BCL-1; BCL-1 oncogene; CCND1; cyclin D1; D11S287E; G1/S-specific cyclin-D1; Kaposi's Sarcoma Virus Type D Cyclin; PRAD1; PRAD1 oncogene; U21B31
Host Species Rat
Immunogen Recombinant fusion protein produced from a clone from KSV.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.