Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KT 182
GREENER_CHOICESDP

Catalog No. 673810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 67-381-0 Supplier Tocris Bioscience™ Supplier No. 6738/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor

KT 182 is a potent, selective and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor (IC50 = 1.7 nM). KT 182 exhibits selectively for ABHD6 over other brain and liver serine hydrolases, but does exhibit some activity at carboxylesterase-1. KT 182 inhibits ABHD6 activity in the brain and liver in mice following intraperitoneal administration.

Specifications

Chemical Name or Material [4-[3 '-(Hydroxymethyl)[1,1 '-biphenyl]-4-yl]-1 H-1,2,3-triazol-1-yl](2-phenyl-1-piperidinyl)methanone
CAS 1402612-62-7
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C27H26N4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.