Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ KT 5823
GREENER_CHOICESDP

Catalog No. 1289100U Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-891-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-891-00U Supplier R&D Systems™ Supplier No. 1289/100U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective protein kinase G inhibitor

Selective inhibitor of protein kinase G (Ki values are 0.23, 4 and > 10 μM for inhibition of PKG, PKC and PKA respectively). Inhibits SNP-stimulated PKG activity with an IC50 of 60 nM in dispersed smooth muscle cells and has little effect on PKA activity at concentrations of up to 10 μM.

Chemical Identifiers

CAS 126643-37-6
Molecular Formula C29H25N3O5

Specifications

Quantity 100 μg
Product Line Tocris
Formula Weight 495.53
Purity 95%
Molecular Weight (g/mol) 495.535
Chemical Name or Material KT 5823
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.