Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab Activated Carbon Filters

Click to view available options
For Use With (Equipment):
Leica Multistainer
Tissue Tek DRS
Description
- Filters contain high grade activated carbon which will absorb most organic and inorganic vapors including xylene, toluene, methanol, formalin, isopropyl alcohol and industrial denatured alcohol
- These products are identical in size and shape to their comparable original manufacturer filters, and are an easy drop-in replacement
- Change filters at manufacturer-recommended intervals to keep your laboratory and staff safe
- 1 Ea.
- Available for Leica MultiStainer or Tissue Tek DRS
Specifications
Specifications
Product Type | Activated Carbon Filter |
For Use With (Equipment) | Leica Multistainer |
Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction