Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab KT5i+ Premium German Advanced Adhesion InkJet Compatible Slides

Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Corner Style:
90°
90° Clipped
Description
- An excellent choice for H and E, special stains, frozen sections and even the toughest IHC procedures with difficult tissues
- These slides feature superior adhesion, maintaining tissue integrity throughout the toughest of tissues or protocols
- Tissue sections easily jump on the slide, indicating a hydrophobic waterbath behavior, but the precision manufacturing process ensures a 100% wettable surface for even reagent spreading
- KT5+i stained slides will not have background staining
- KT5+i slides provide a colored marking area for efficient specimen color-coding, identification, labeling and printing
- KT5+i slides are available in 5 unique colors, with 45° or 90° corners
- KT5+i Slides are compatible with both inkjet and UV laser printers
- Manufactured from premium German glass that enables superior clarity for digital pathology and imaging systems
- Dimenstions: 75.6 x 25.6 x 1 mm
- 10 gross per case.
Specifications
Specifications
| Length (Metric) | 75.6 mm |
| Precleaned | Yes |
| Material | High Quality Glass |
| Thickness (Metric) | 1 mm |
| Type | Adhesion Slide |
| Width (Metric) | 25.6 mm |
| Sterility | Non-sterile |
| Edges | 90°, Ground |
| Frosted | 1 End, 1 Side |
| Well Count | 0 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction