Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KT3 Epitope Tag Antibody, DyLight 405, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB11040654V
Description
KT3 Epitope Tag Polyclonal antibody specifically detects KT3 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
KT3 Epitope Tag | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
KPPTPPPEPET Epitope Tag, KPPTPPPEPET Tag, KT3 Epitope Tag | |
Goats were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction