Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KTI12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545752 Supplier Invitrogen™ Supplier No. PA545752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VPKELEREES GAAESPALVT PDSEKSAKHG SGAFYSPELL EALTLRFEAP Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KTI12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KTI12
Gene Accession No. Q96EK9
Gene Alias KTI12; KTI12 chromatin associated homolog; KTI12 homolog, chromatin associated; KTI12 homolog, chromatin associated (S. cerevisiae); protein KTI12 homolog; RP11-91A18.3; SBBI81; TOT4
Gene Symbols KTI12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KTI12.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112970
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.