Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KTI12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556670
Description
Immunogen sequence: PLCLVYCVRP GGPIAGPQVA GANENPGRNV SVSWRPRAEE DGRAQAAGSS VLRELHTADS VVNGSAQADV PKELEREESG Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 31%.
The function of this protein remains unknown.
Specifications
KTI12 | |
Polyclonal | |
Unconjugated | |
KTI12 | |
KTI12; KTI12 chromatin associated homolog; KTI12 homolog, chromatin associated; KTI12 homolog, chromatin associated (S. cerevisiae); protein KTI12 homolog; RP11-91A18.3; SBBI81; TOT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EK9 | |
KTI12 | |
Recombinant protein corresponding to Human KTI12. Recombinant protein control fragment (Product #RP-94554). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction