Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KTI12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556671 Supplier Thermo Scientific Supplier No. PA556671
Add to Cart
Add to Cart

KTI12 Polyclonal Antibody

KTI12 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen KTI12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KTI12
Gene Accession No. Q96EK9
Gene Alias KTI12; KTI12 chromatin associated homolog; KTI12 homolog, chromatin associated; KTI12 homolog, chromatin associated (S. cerevisiae); protein KTI12 homolog; RP11-91A18.3; SBBI81; TOT4
Gene Symbols KTI12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KTI12. Recombinant protein control fragment (Product #RP-94555).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.