Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KTI12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598683
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598683 Supplier Invitrogen™ Supplier No. PA598683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KTI12 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KTI12
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KTI12
Gene Accession No. Q5I0L7, Q96EK9
Gene Alias KTI12; KTI12 chromatin associated homolog; KTI12 homolog, chromatin associated; KTI12 homolog, chromatin associated (S. cerevisiae); protein KTI12 homolog; RP11-91A18.3; SBBI81; TOT4
Gene Symbols KTI12
Host Species Rabbit
Immunogen Recombinant Human Protein KTI12 homolog protein (110-204AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112970, 685656
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.