Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553652 Supplier Invitrogen™ Supplier No. PA553652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELKRLEAMLK ERESDLSSKT QLLQDVQDEN KLFKSQIEQL KQQNYQQASS FPPHEELLKV ISEREKEISG LWNELDSLKD AVEHQRKKNN ERQQQVEAVE LEAKE Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.

Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KTN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KTN1
Gene Accession No. Q86UP2
Gene Alias CG1; CG-1 antigen; KIAA0004; Kinectin; kinectin 1; kinectin 1 (kinesin receptor); kinesin receptor; KNT; ktn1; MU-RMS-40.19
Gene Symbols KTN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KTN1. Recombinant protein control fragment (Product #RP-89002).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.