Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588557 Supplier Invitrogen™ Supplier No. PA588557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEFYESAYFI VLIPSIVITV IFLFFWLFMK ETLYDEVLAK QKREQKLIPT KTDKKKAEKK KNKKKEIQNG NLHESDSESV PRDFKLSDAL AVEDDQVAPV; Positive Samples: K-562, SKOV3, Raji, HeLa, HT-1080, Mouse heart, Rat testis; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.

Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KTN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KTN1
Gene Accession No. Q61595, Q86UP2
Gene Alias CG1; CG-1 antigen; KIAA0004; Kinectin; kinectin 1; kinectin 1 (kinesin receptor); kinesin receptor; KNT; ktn1; MU-RMS-40.19
Gene Symbols KTN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KTN1 (NP_0010729891).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16709, 361029, 3895
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.