Learn More
KTN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | KTN1 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2566
![]() ![]() |
Proteintech Group Inc
198411AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.Specifications
KTN1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q61595, Q86UP2 | |
16709, 3895 | |
KTN1 Fusion Protein Ag13854 | |
Primary | |
-20°C | |
KTN1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CG 1 antigen, CG1, KIAA0004, Kinectin, kinectin 1 (kinesin receptor), Kinesin receptor, KNT, KTN1, MU RMS 40.19 | |
KTN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.