Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KU 0060648

Catalog No. 484010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-401-0 10 mg
48-405-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-401-0 Supplier Tocris Bioscience™ Supplier No. 4840/10
This item is not returnable. View return policy

Dual DNA-PK and PI 3-K inhibitor

Dual PI 3-K and DNA-PK inhibitor (IC50 values are <0.1, 0.5, 4 and 19 nM for PI 3-Kδ, PI 3-Kβ, PI 3-Kα and DNA-PK respectively). Inhibits proliferation of MCF7 cells in vitro and delays growth of MCF7 xenografts in mice. Also enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency ∽2 to 4-fold, and decreases nonhomologous end-joining (NHEJ) frequency ∽40%; maximum effect seen at 250 nM.

Chemical Identifiers

CAS 881375-00-4
Molecular Formula C33H34N4O4S

Specifications

Quantity 10 mg
Formula Weight 582.71
Purity >97%
Molecular Weight (g/mol) 582.719
Chemical Name or Material KU 0060648
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.