Learn More
Invitrogen™ Ku70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120056
Description
Positive test controls include: SKOV3, COS7, SW480, HeLa, Mouse brain, Mouse liver, Mouse skeletal muscle, Rat liver. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: PYFVALVPQE EELDDQKIQV TPPGFQLVFL PFADDKRKMP FTEKIMATPE QVGKMKAIVE KLRFTYRSDS FENPVLQQHF RNLEALALDL MEPEQAVDLT LPKVEAMNKR LGSLVDEFKE LVYPPDYNPE GKVTKRKHDN EGSGSKRPKV EYSEEELKTH ISKGTLGKFT VPMLKEACRA YGLKSGLKKQ ELLEALTKHF QD.
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.
Specifications
Ku70 | |
Polyclonal | |
Unconjugated | |
XRCC6 | |
5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 70kDa; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CT; CTA-216E10.7; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTCBF; DNA repair protein XRCC6; G22p1; Ku autoantigen p70 subunit; ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Kup70; Lupus Ku autoantigen protein p70; ML8; OTTHUMP00000028581; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; XELAEV_18023498mg; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair complementing defective repair in Chinese hamster cells 6 L homeolog; X-ray repair cross complementing 6; X-ray repair cross-complementing protein 6; xrcc6; xrcc6.L | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14375, 25019, 2547 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12956, P23475 | |
XRCC6 | |
A synthetic peptide corresponding to a sequence within amino acids 408-609 of human Ku70 (NP_0014601). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.