Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ku70/XRCC6 Antibody (7A9E7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Ku70/XRCC6 |
---|---|
Clone | 7A9E7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
Description
Ku70/XRCC6 Monoclonal specifically detects Ku70/XRCC6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
Ku70/XRCC6 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
PBS with 0.05% Sodium Azide | |
2547 | |
Purified recombinant fragment of human XRCC6 (AA: 6-214) expressed in E. Coli. | |
Primary | |
69.8 kDa |
7A9E7 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6 | |
XRCC6 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title