Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ku70/XRCC6 Antibody (KU729), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234663AF405
Description
Ku70/XRCC6 Monoclonal specifically detects Ku70/XRCC6 in Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
Ku70/XRCC6 | |
Monoclonal | |
Alexa Fluor 405 | |
5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6 | |
Mouse | |
75 kDa | |
0.1 mL | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
2547 | |
Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
KU729 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
XRCC6 | |
Nuclear extract of human HL-60 cells | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a dimer of two proteins of 70kDa and ∼80kDa, identified as two subunits of Ku. This monoclonal antibody recognizes a conformational epitope of p70/p80 dimer, which is destroyed during Western blotting. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction