Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ku80 Monoclonal Antibody (149.8)
GREENER_CHOICE

Catalog No. PIMA531593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531593 Supplier Invitrogen™ Supplier No. MA531593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: Jurkat, Raji, 293T, A431, HeLa, HepG2, H1299, HCT116, MCF-7, NT2D1, PC-3, U87-MG.

The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ku80
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 149.8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene XRCC5
Gene Accession No. P13010
Gene Alias 86 kDa subunit of Ku antigen; AI314015; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; FLJ39089; G22p2; HGNC:12833; KARP1; KARP-1; Ku autoantigen; ku autoantigen protein p86 homolog; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p80; Ku80; Ku-80; Ku86; Ku-86; Ku86 autoantigen related protein 1; KUB2; Kup80; Lupu; lupus; lupus Ku autoantigen protein p86; NFIV; nuclear factor IV; OTTHUMP00000164061; OTTHUMP00000206791; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining; Ku autoantigen, 80kDa); X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break rejoining; Ku autoantigen, 80kD); X-ray repair cross complementing 5; X-ray repair cross-complementing protein 5; Xrcc5
Gene Symbols XRCC5
Host Species Mouse
Immunogen Recombinant antigen containing the carboxy terminal portion of human Ku80 protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7520
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.