Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ku80/XRCC5 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen Ku80/XRCC5
Concentration 1 mg/mL
Dilution Western Blot 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Applications Western Blot, Immunoprecipitation, ChIP Assay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100508SS
SDP
View Documents
Novus Biologicals
NB100508SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100508
SDP
View Documents
Novus Biologicals
NB100508
0.1 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Ku80/XRCC5 Polyclonal specifically detects Ku80/XRCC5 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Ku80/XRCC5
Western Blot 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Polyclonal
Rabbit
Cancer, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Stem Cell Markers
PBS with 0.02% Sodium Azide
7520
A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, Immunoprecipitation, ChIP Assay
Unconjugated
RUO
Human, Mouse, Rat, Hamster
80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster
XRCC5
IgG
Affinity Purified
Reacts with residues 419-440 [LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein. Sequence is 100% conserved between human, mouse, rat and hamster.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.