Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kudoa thyrsites Monoclonal Antibody (IPA-2F4)
GREENER_CHOICE

Catalog No. PIMA170114
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA170114 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA170114 Supplier Invitrogen™ Supplier No. MA170114
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-70114 detects Kudoa Thrysites in Kudoa thrysites samples. MA1-70114 has been successfully used in Western blot, immunofluorescence and flow cytometry procedures. The MA1-70114 immunogen is PercollTM-purified K. thyrsites spores.

Kudoa thrysites is a myxosporean parasite of marine fishes. It has a worldwide distribution, and infects a wide range of host species. This parasite is responsible for causing economic losses to the fisheries sector, by causing post-mortem myoliquefaction, a softening of the flesh to such an extent that the fish becomes unmarketable. It is not infective to humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kudoa thyrsites
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone IPA-2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias myxosporean
Host Species Mouse
Immunogen PercollTM-purified K. thyrsites spores.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.