Learn More
PIP™ Kut-Gard™ Polyester-Wrapped Stainless Steel Core Cut-Resistant Gloves
Shop All Protective Industrial Products Inc. ProductsDescription
- Seamless knit construction provides comfort, fit and dexterity
- PolyKor™ engineered yarns offer cut and slash resistance
- Ambidextrous pattern permits glove to be worn on either hand
- Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Secure gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Products constructed with PolyKor™ engineered yarn are suitable for protection against mechanical hazards in the toughest environments
- PolyKor products are highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR, 170-199 |
| Construction | Seamless Knit |
| Coating Material | Uncoated |
| Color | Blue |
| Cut Resistance Rating | ANSI A5 |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Stainless Steel Fiber, Synthetic Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.