Learn More
Invitrogen™ KV1.1 (KCNA1) Polyclonal Antibody

Description
Antibody detects endogenous levels of total KCNA1.
Specifications
Specifications
| Antigen | KV1.1 (KCNA1) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Kcna1 |
| Gene Accession No. | P10499, P16388, Q09470 |
| Gene Alias | AEMK; AI840627; brain potassium channel protein-1; EA1; HBK1; HUK1; IA; Kca1-1; Kcna; Kcna1; Kcpvd; Kv1.1; MBK1; mceph; megencephaly; MGC126782; MGC138385; MK1; Mk-1; MKI; potassium (K+) channel protein voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 1; potassium voltage gated channel shaker related subfamily member 1; potassium voltage gated channel, shaker related subfamily, member 1; potassium voltage-gated channel subfamily A member 1; potassium voltage-gated channel, shaker-related subfamily, member 1; potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia); RBK1; RBKI; RCK1; Shak; Voltage-gated K(+) channel HuKI; voltage-gated potassium channel HBK1; Voltage-gated potassium channel subunit Kv1.1 |
| Gene Symbols | Kcna1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.