Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV1.2 (KCNA2) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144086
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144086 Supplier Invitrogen™ Supplier No. PA5144086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is identical to the related mouse and rat sequences. Positive Control - WB: Rat Kidney Tissue, Rat Brain Tissue, Mouse Brain Tissue, Mouse Kidney Tissue. ICC/IF: U20S cell. Flow: A431 cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Kcna2 is a voltage-gated potassium channel (KV) that belongs to the 6-TM family of potassium channel and also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). The Kcna2 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. Kcna2 contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. Further, Kcna2 belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. The coding region of the Kcna2 gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1. Diseases associated with KCNA2 include Epileptic Encephalopathy (Early Infantile, 32) and Undetermined Early-Onset Epileptic Encephalopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.2 (KCNA2)
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene KCNA2
Gene Accession No. P16389, P63141, P63142
Gene Alias Akr6a4; BK2; CSMK1; delayed rectifier K+ channel; EIEE32; ENSMUSG00000074335; Gm10672; HBK5; HK4; HUKIV; k(v)1.2; KC22; Kca1-2; kcna2; kcna2.S; kcna2-a; KV1.2; Kv1.2' potassium channel; LOC100537815; MK2; Mk-2; NGK1; Potassium (K+) channel protein alpha 2, voltage dependent; potassium channel (BGK5); potassium channel subunit Kv 1.2; potassium channel, voltage gated shaker related subfamily A, member 2; potassium channel, voltage gated shaker related subfamily A, member 2 S homeolog; potassium voltage gated channel shaker related subfamily member 2; potassium voltage-gated channel subfamily A member 2; potassium voltage-gated channel, shaker-related subfamily, member 2; potassium voltage-gated channel, shaker-related subfamily, member 2 a; RAK; RBK2; RCK5; RP11-284N8.1; voltage-dependent K channel; voltage-gated channel, shaker-related subfamily, member 2; voltage-gated K(+) channel HuKIV; Voltage-gated potassium channel HBK5; voltage-gated potassium channel isoform 2; voltage-gated potassium channel isoform 3; voltage-gated potassium channel protein Kv1.2; Voltage-gated potassium channel subunit Kv1.2; XELAEV_18011981mg; XSha2
Gene Symbols KCNA2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Kv1.2 (466-499aa NNSNEDFREENLKTANCTLANTNYVNITKMLTDV).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16490, 25468, 3737
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.