Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv1.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | Kv1.2 |
---|---|
Dilution | Western Blot 5 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Kv1.2 Polyclonal antibody specifically detects Kv1.2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Kv1.2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Neuroscience, Neurotransmission, Potassium Channels | |
PBS | |
3737 | |
IgG | |
Ammonium sulfate precipitation |
Western Blot 5 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.6.1.27, EC 6.1.1, HBK5, HK4, HUKIV, Kv1.2, MGC50217, MK2, NGK1, potassium channel, potassium voltage-gated channel subfamily A member 2, potassium voltage-gated channel, shaker-related subfamily, member 2, RBK2, Voltage-gated K(+) channel HuKIV, Voltage-gated potassium channel HBK5, voltage-gated potassium channel protein Kv1.2, Voltage-gated potassium channel subunit Kv1.2 | |
Synthetic peptide derived from the rat Kv1.2 potassium channel conjugated to KLH | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title