Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KV1.3 (KCNA3) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577571
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications
KV1.3 (KCNA3) | |
Polyclonal | |
Unconjugated | |
KCNA3 | |
Kcna3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16491, 29731, 3738 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 1% BSA, 5% sucrose and 0.05% sodium azide; pH 7.4 | |
P15384, P16390, P22001 | |
Kcna3 | |
GST fusion protein with sequence TLSKSEYMVIEEGGMNHSAFPQTPFKTGNSTATCTTNNNPNSCVNIKKIFTDV, corresponding to amino acid residues 523-575 of human Kv 1.3 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction