Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV1.4 (KCNA4) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553416 Supplier Invitrogen™ Supplier No. PA553416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFNYFYHRET ENEEQTQLTQ NAVSCPYLPS NLLKKFRSST SSSLGDKSEY LEMEEGVKES LCAKEEKCQG KGDDSETDKN NCSNAKAVET DV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

Potassium voltage-gated channel subfamily A member 4, also known as Kv1.4 or PCN2, is a protein that in humans is encoded by the KCNA4 gene. This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. It is mapped to 11p14.1. KCNA4 belongs to the A-type potassium current class, the members of which may be important in the regulation of the fast repolarizing phase of action potentials in heart and thus may influence the duration of cardiac action potential. KCNA4 also contributes to the cardiac transient outward potassium current (Ito1), the main contributing current to the repolarizing phase 1 of the cardiac action potential. This gene has been shown to interact with DLG4, KCNA2 and DLG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.4 (KCNA4)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNA4
Gene Accession No. P22459
Gene Alias cardiac potassium channel; fetal skeletal muscle potassium channel; HBK4; HK1; HPCN2; HUKII; KCHAN; Kcna4; KCNA4L; KCNA8; Kv1.4; Kv4; PCN2; potassium channel 2; potassium channel, voltage gated shaker related subfamily A, member 4; potassium voltage gated channel, shaker related subfamily, member 4; potassium voltage-gated channel subfamily A member 4; potassium voltage-gated channel, shaker-related subfamily, member 4; rapidly inactivating potassium channel; RCK4; RHK1; RK3; shaker-related potassium channel Kv1.4; type A potassium channel; voltage-gated K(+) channel HuKII; Voltage-gated potassium channel HBK4; voltage-gated potassium channel HK1; Voltage-gated potassium channel subunit Kv1.4
Gene Symbols KCNA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNA4. Recombinant protein control fragment (Product #RP-91125).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.