Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv1.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | Kv1.4 |
---|---|
Dilution | Western Blot 5 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Kv1.4 Polyclonal antibody specifically detects Kv1.4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Kv1.4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
cardiac potassium channel, fetal skeletal muscle potassium channel, HBK4, HK1, HPCN2HUKII, KCNA4L, KCNA8, Kv1.4, PCN2, potassium channel 2, potassium voltage-gated channel subfamily A member 4, potassium voltage-gated channel, shaker-related subfamily, member 4, potassium voltage-gated channel, shaker-related subfamily, member 4-like, rapidly inactivating potassium channel, shaker-related potassium channel Kv1.4, type A potassium channel, Voltage-gated K(+) channel HuKII, Voltage-gated potassium channel HBK4, Voltage-gated potassium channel HK1, Voltage-gated potassium channel subunit Kv1.4 | |
Synthetic peptide derived from the N-terminus of the rat Kv1.4 potassium channel conjugated to KLH (Uniprot: P22459) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 5 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
3739 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title