Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv1.4 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094668 20 μL
NB094667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094668 Supplier Novus Biologicals Supplier No. NBP30298020UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv1.4 Polyclonal specifically detects Kv1.4 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Kv1.4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias cardiac potassium channel, fetal skeletal muscle potassium channel, HBK4, HK1, HPCN2HUKII, KCNA4L, KCNA8, Kv1.4, PCN2, potassium channel 2, potassium voltage-gated channel subfamily A member 4, potassium voltage-gated channel, shaker-related subfamily, member 4, potassium voltage-gated channel, shaker-related subfamily, member 4-like, rapidly inactivating potassium channel, shaker-related potassium channel Kv1.4, type A potassium channel, Voltage-gated K(+) channel HuKII, Voltage-gated potassium channel HBK4, Voltage-gated potassium channel HK1, Voltage-gated potassium channel subunit Kv1.4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Kv1.4 (NP_002224.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3739
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.