Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV1.5 (KCNA5) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540815 Supplier Invitrogen™ Supplier No. PA540815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DPGVRPLPPL PEELPRPRRP PPEDEEEEGD PGLGTVEDQA LGTASLHHQR Sequence homology: Human: 100%; Mouse: 93%; Rat: 93%.

In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.5 (KCNA5)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kcna5
Gene Accession No. P22460
Gene Alias ATFB7; cardiac potassium channel; delayed rectifier K+ channel; HCK1; HK2; HPCN1; insulinoma and islet potassium channel; KCNA5; Kv1; KV1.5; KV1-5; MGC117058; MGC117059; PCN1; Potassium (K+) channel protein alpha 5; potassium channel 1; potassium channel, voltage gated shaker related subfamily A, member 5; potassium voltage gated channel, shaker related subfamily, member 5; potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; voltage-gated potassium channel HK2; voltage-gated potassium channel protein Kv1.5; voltage-gated potassium channel subunit Kv1.5
Gene Symbols Kcna5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCNA5 (aa 72-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3741
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.