Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV1.7 (KCNA7) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541062 Supplier Invitrogen™ Supplier No. PA541062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GEEAGMFSHV DMQPCGPLEG KANGGLVDGE VPELPPPLWA PPGKHLVTEV Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 100%.

Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.7 (KCNA7)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNA7
Gene Accession No. Q96RP8
Gene Alias HAK6; Kcna7; Kcnc7; Kv1.7; potassium channel, voltage gated shaker related subfamily A, member 7; potassium voltage gated channel, shaker related subfamily, member 7; potassium voltage-gated channel subfamily A member 7; potassium voltage-gated channel, shaker-related subfamily, member 7; potassium voltage-gated channel, shaker-related subfamily, member 7; potassium voltage gated channel, shaker related subfamily, member 7; voltage-dependent potassium channel Kv1.7; voltage-gated potassium channel KCNA7; voltage-gated potassium channel subunit Kv1.7
Gene Symbols KCNA7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KCNA7 (aa 407-456).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3743
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.