Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv11.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2841260.1ML
Description
Kv11.2 Polyclonal specifically detects Kv11.2 in Human samples. It is validated for Western Blot.Specifications
Kv11.2 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
81033 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
eag related protein 2, eag-related gene member 2, Eag-related protein 2, erg2, ERG-2, ether-a-go-go related gene potassium channel 2, Ether-a-go-go-related gene potassium channel 2, Ether-a-go-go-related protein 2, hERG-2, HERG2, Kv11.2, potassium voltage-gated channel subfamily H member 6, potassium voltage-gated channel, subfamily H (eag-related), member 6, Voltage-gated potassium channel subunit Kv11.2 | |
The immunogen is a synthetic peptide directed towards the middle region of human Kv11.2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction