Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv12.1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32141125UL
Description
Kv12.1 Polyclonal antibody specifically detects Kv12.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Kv12.1 | |
Polyclonal | |
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
ELK, ELK channel 3, ELK1, ELK3, ether-a-go-go-like potassium channel 1, Ether-a-go-go-like potassium channel 3, hElk1, Kv12.1, potassium voltage-gated channel subfamily H member 8, potassium voltage-gated channel, subfamily H (eag-related), member 8, Voltage-gated potassium channel subunit Kv12.1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SEVTTLTQEVSQLGKDMRNVIQLLENVLSPQQPSRFCSLHSTSVCPSRESLQTRTSWSAHQPCLHLQTGGAAYTQAQLCSSNITSDIWSVDPSSVGSSPQRTGAHEQNPADSELYHSPSLDYSPSHYQVVQEGHL | |
25 μg | |
Signal Transduction | |
131096 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction