Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KV2.1 (KCNB1) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579551
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. IHC: human lung cancer tissue, rat brain tissue, mouse brain tissue IHC-F: rat brain tissue, mouse brain tissue.
Mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient.
Specifications
KV2.1 (KCNB1) | |
Polyclonal | |
Unconjugated | |
Kcnb1 | |
Delayed rectifier potassium channel 1; delayed rectifier potassium channel Kv2.1; DRK1; DRK1PC; KCB1; Kcnb1; Kcr1-1; Kv2.1; mShab; potassium channel Kv2.1; potassium channel, voltage gated Shab-related subfamily B, member 1; potassium voltage gated channel, Shab-related subfamily, member 1; potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Shab; voltage-gated potassium channel subunit Kv2.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16500, 25736, 3745 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P15387, Q03717, Q14721 | |
Kcnb1 | |
E.coli-derived human Kv2.1 recombinant protein (Position: V687-I858). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction