Learn More
KV2.1 (KCNB1) Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111830
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)HMLPGGG AHGSTRDQSI Homology: Mouse amino acid residues identical; human - 15/17 amino acid residues identical.
Mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient.
Specifications
KV2.1 (KCNB1) | |
Polyclonal | |
Unconjugated | |
Kcnb1 | |
BOS_13247; Delayed rectifier potassium channel 1; DRK1; DRK1PC; KCB1; KCNB1; Kcr1-1; Kv2.1; mShab; Potassium voltage-gated channel subfamily B member 1; Shab; Voltage-gated potassium channel subunit Kv2.1 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
16500, 25736 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P15387, Q03717 | |
Kcnb1 | |
Peptide corresponding to amino acid residues 841-857 of rat KV2.1 (Intracellular, C-terminus) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.