Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV2.2 (KCNB2) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566815 Supplier Invitrogen™ Supplier No. PA566815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHISTILLEET PSQGDRPLLG TEVSAPCQGP SKGLSPRFPK QKLFPFSSRE RRSFTEIDTG DDEDFLELP Highest antigen sequence identity to the following orthologs - mouse 79%, rat 81%.

Kcnb2 (Kv2.2) encodes a Voltage-gated K+ channel which are important determinants of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variation in action potential waveforms and repetitive firing patterns evident in different neuronal cell types. The delayed rectifier-type (IK) channels (Kv1.5, Kv2.1, and Kv2.2) are expressed on all neuronal somata and proximal dendrites and are also found in a wide variety on non-neuronal cell types including pancreatic islet, alveolar cells and cardiac myocytes. Kv2.1 and Kcnb2 form distinct populations of K+ channels and these subunits are thought to be primarily responsible for IK in superior cervical ganglion cells. Functionally, Kcnb2 mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient. Diseases associated with KCNB2 include Brugada Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV2.2 (KCNB2)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNB2
Gene Accession No. Q92953
Gene Alias 9630047L19Rik; BB130875; CDRK; delayed rectifier potassium channel protein; kcnb2; kcnb2.L; kcnb2-a; kcnb2-b; Kv2.2; potassium channel Kv2.2; potassium channel, voltage gated Shab related subfamily B, member 2; potassium channel, voltage gated Shab related subfamily B, member 2 L homeolog; potassium channel, voltage gated Shab-related subfamily B, member 2; potassium voltage gated channel, Shab-related subfamily, member 2; potassium voltage-gated channel subfamily B member 2; potassium voltage-gated channel, Shab-related subfamily, member 2; voltage-gated potassium channel subunit Kv2.2
Gene Symbols KCNB2
Host Species Rabbit
Immunogen Recombinant Human KCNB2. Recombinant protein control fragment (Product #RP-107471).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.