Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KV3.2 (KCNC2) Mouse anti-Human, Mouse, Rat, Clone: S410-17, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527683
Description
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels. The subunits vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits, which include the KV1, KV2, KV3, KV4 and KV9 proteins, as well as accessory subunits that modify the gating properties of the coexpressed KV subunits. KV3.2 is a multipass membrane protein that regulates the voltage-dependent K+ permeability of excitable membranes. The tail of KV3.2 may be influential in the targeting of the channel to specific subcellular compartments and/or the regulation of channel activity.
Specifications
KV3.2 (KCNC2) | |
Monoclonal | |
1 mg/ml | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 | |
P22462, Q14B80, Q96PR1 | |
Kcnc2 | |
Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
S410-17 | |
Unconjugated | |
KCNC2 | |
Potassium voltage-gated channel subfamily C member 2, Kcnc2, KSHIIIA | |
Mouse | |
Protein G | |
RUO | |
246153, 268345, 3747 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction