Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV3.2 (KCNC2) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115448 Supplier Thermo Scientific Supplier No. PA5115448
Add to Cart
Add to Cart

KV3.2 (KCNC2) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen KV3.2 (KCNC2)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene KCNC2
Gene Accession No. P22462, Q14B80, Q96PR1
Gene Alias AW047325; B230117I07; KCNC2; KShIIIA; Kv3.2; Potassium channel voltage-gated Shaw-related subfamily C member 2; potassium channel, voltage gated Shaw related subfamily C, member 2; potassium channel, voltage gated Shaw-related subfamily C, member 2; potassium voltage gated channel, Shaw-related subfamily, member 2; potassium voltage-gated channel subfamily C member 2; potassium voltage-gated channel, Shaw-related subfamily, member 2; shaw-like potassium channel; Voltage-gated potassium channel Kv3.2; voltage-gated potassium channel subunit Kv3.2
Gene Symbols KCNC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNC2(Accession Q96PR1), corresponding to amino acid residues G180-F230.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246153, 268345, 3747
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.