Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.2 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: N410/17, MilliporeSigma™

Catalog No. MABN24321MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN24321MI 100 μg
MABN24322MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN24321MI Supplier MilliporeSigma Supplier No. MABN2432100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kv3.2 Monoclonal antibody specifically detects Kv3.2 in Human,Mouse,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv3.2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N410/17
Conjugate Unconjugated
Gene Accession No. NP_631962
Gene Alias Potassium voltage-gated channel subfamily C member 2;Potassium channel voltage-gated Shaw-related subfamily C member 2;Shaw-like potassium channel;Voltage-gated potassium channel subunit Kv3.2.
Gene Symbols Kcnc2
Host Species Mouse
Immunogen A recombinant fragment corresponding to 140 amino acids from the C-terminal region of rat Voltage-gated potassium channel subunit Kv3.2.
Molecular Weight of Antigen ∼67 kDa observed; 67.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone N410/17 is a mouse monoclonal antibody that specifically detects Voltage-gated potassium channel subunit Kv3.2. It targets an epitope with in the C-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.