Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KV3.3 (KCNC3) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115449
Description
Antibody detects endogenous levels of total KCNC3.
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes.
Specifications
KV3.3 (KCNC3) | |
Polyclonal | |
Unconjugated | |
KCNC3 | |
KCNC3; Kcr2-3; KShIIID; KV3.3; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily C member 3; potassium channel, voltage gated Shaw related subfamily C, member 3; potassium channel, voltage gated Shaw-related subfamily C, member 3; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; SCA13; Shaw type potassium channel Kv3.3; Shaw-related voltage-gated potassium channel protein 3; voltage-gated potassium channel protein KV3.3; voltage-gated potassium channel subunit Kv3.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117101, 16504, 3748 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q01956, Q14003, Q63959 | |
KCNC3 | |
A synthesized peptide derived from human KCNC3(Accession Q14003), corresponding to amino acid residues A671-I721. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction