Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV3.4 (KCNC4) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553226 Supplier Invitrogen™ Supplier No. PA553226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISSVCVSSYR GRKSGNKPPS KTCLKEEMAK GEASEKIIIN VGGTRHETYR STLRTLPGTR LAWLADPD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 96%.

The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. It generates atypical voltage-dependent transient current that may be important for neuronal excitability. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV3.4 (KCNC4)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNC4
Gene Accession No. Q03721
Gene Alias AI850292; C1orf30; HKSHIIIC; K+ channel subunit; KCNC4; Kcr2-4; KSHIIIC; Kv3.4; MGC126818; potassium channel, voltage gated Shaw related subfamily C, member 4; potassium channel, voltage gated Shaw-related subfamily C, member 4; potassium voltage gated channel, Shaw-related subfamily, member 4; potassium voltage-gated channel subfamily C member 4; potassium voltage-gated channel subfamily C member 4 (Voltage-gated potassium channel subunit Kv3.4) (Raw3); potassium voltage-gated channel, Shaw-related subfamily, member 4; raw3; Voltage-gated potassium channel subunit Kv3.4
Gene Symbols KCNC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNC4. Recombinant protein control fragment (Product #RP-91089).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3749
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.