Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV3.4 (KCNC4) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590618 Supplier Invitrogen™ Supplier No. PA590618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKRKKHVPRP AQLESPMYCK SEETSPRDST CSDTSPPARE EGMIERKRA; Positive Samples: U-87MG, Mouse brain; Cellular Location: Membrane, Multi-pass membrane protein.

The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. It generates atypical voltage-dependent transient current that may be important for neuronal excitability. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV3.4 (KCNC4)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNC4
Gene Accession No. Q03721, Q8R1C0
Gene Alias AI850292; C1orf30; HKSHIIIC; K+ channel subunit; KCNC4; Kcr2-4; KSHIIIC; Kv3.4; MGC126818; potassium channel, voltage gated Shaw related subfamily C, member 4; potassium channel, voltage gated Shaw-related subfamily C, member 4; potassium voltage gated channel, Shaw-related subfamily, member 4; potassium voltage-gated channel subfamily C member 4; potassium voltage-gated channel subfamily C member 4 (Voltage-gated potassium channel subunit Kv3.4) (Raw3); potassium voltage-gated channel, Shaw-related subfamily, member 4; raw3; Voltage-gated potassium channel subunit Kv3.4
Gene Symbols KCNC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 490-538 of human KCNC4 (NP_0049692).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3749, 99738
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.