Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv3.4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2851890.1ML
Description
Kv3.4 Polyclonal specifically detects Kv3.4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kv3.4 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
3749 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4 | |
The immunogen is a synthetic peptide directed towards the middle region of human Kv3.4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction