Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv4.1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25512825UL
Description
Kv4.1 Polyclonal specifically detects Kv4.1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Kv4.1 | |
Polyclonal | |
Western Blot 1:100 - 1:250, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
Kv4.1, potassium voltage-gated channel subfamily D member 1, potassium voltage-gated channel, Shal-related subfamily, member 1, shal-type potassium channel, voltage-gated potassium channel Kv4.1, Voltage-gated potassium channel subunit Kv4.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
3750 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
KCND1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NTLDRYPDTLLGSSEKEFFYDADSGEYFFDRDPDMFRHVLNFYRTGRLHCPRQECIQAFDEELAFYGLVPELVGDCCLEEYRDRKKENAERLAEDEEAEQ | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction