Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv4.2 Antibody (5B11B9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Kv4.2 |
---|---|
Clone | 5B11B9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
Kv4.2 Monoclonal specifically detects Kv4.2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Kv4.2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS with 0.05% Sodium Azide | |
3751 | |
Purified recombinant fragment of human KCND2 (AA: 27-184) expressed in E. Coli. | |
Primary | |
70.5 kDa |
5B11B9 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
KCND2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title