Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV4.3 (KCND3) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593292 Supplier Invitrogen™ Supplier No. PA593292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESSMQNYPS TRSPSLSSHP GLTTTCCSRR SKKTTHLPNS NLPATRLRSM QELSTIHIQG SEQPSLTTSR SSLNLKADDG LRPNCKTSQI TTAIISIPTP PALTPEGESR PPPASPGPNT NIPSIASNVV KVSAL; Positive Samples: Mouse liver, Mouse brain, Rat liver; Cellular Location: Cell membrane, Cell projection, Membrane, Multi-pass membrane protein, dendrite, sarcolemma.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential. This member includes two isoforms with different sizes, which are encoded by alternatively spliced transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV4.3 (KCND3)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 8.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KCND3
Gene Accession No. Q62897, Q9UK17, Q9Z0V1
Gene Alias AW045978; BRGDA9; KCND3; KCND3L; KCND3S; Kncd3; KSHIVB; KV4.3; potassium channel alpha subunit Kv4.3 short form; potassium channel Kv4.3; potassium channel Kv4.3L; potassium channel Kv4.3M; potassium channel, voltage gated Shal related subfamily D, member 3; potassium channel, voltage-gated Shal-related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage gated channel, Shal-related family, member 3; potassium voltage-gated channel long; potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related family, member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; SCA19; SCA22; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; voltage-gated potassium channel subunit Kv4.3
Gene Symbols KCND3
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3752, 56543, 65195
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.