Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv7.5 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298067PCP
Description
Kv7.5 Polyclonal antibody specifically detects Kv7.5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Kv7.5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
KQT-like 5, Kv7.5, potassium channel protein, Potassium channel subunit alpha KvLQT5, potassium voltage-gated channel subfamily KQT member 5, potassium voltage-gated channel, KQT-like subfamily, member 5, Voltage-gated potassium channel subunit Kv7.5 | |
Produced in rabbits immunized with E. coli-derived Human Kv7.5 fragment. | |
0.1 mL | |
Signal Transduction | |
56479 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction